Methyl-4-chlorsalicylat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-chloro-2-hydroxybenzoate

CA-4473-5G CA-4473-25G CA-4473-1G
COBBCA-4473-5GEA 114 EUR
COBBCA-4473-5G COBBCA-4473-25G COBBCA-4473-1G
Methyl-4-chlorsalicylat
Methyl-4-chlorsalicylat
Formel: C₈H₇ClO₃
Molecular Weight: 186,5900 g/mol
MDL: MFCD03407436
CAS-Nummer: 22717-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR