Methyl-6-fluorsalicylat
Lieferant: FLUOROCHEM
Synonyme:
Methyl 2-fluoro-6-hydroxybenzoate
006803-1G
006803-2.5G
FLUO006803-1GEA
122
EUR
FLUO006803-1G
FLUO006803-2.5G
Methyl-6-fluorsalicylat
Methyl-6-fluorsalicylat
Formel:
C₈H₇FO₃ Molecular Weight: 170,1378 g/mol |
MDL:
MFCD00153207 CAS-Nummer: 72373-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...