4'-Nitro-2'-(trifluormethyl)acetanilid

Lieferant: FLUOROCHEM

006901-5G 006901-25G
FLUO006901-5GEA 57.9 EUR
FLUO006901-5G FLUO006901-25G
4'-Nitro-2'-(trifluormethyl)acetanilid
4'-Nitro-2'-(trifluormethyl)acetanilid
Formel: C₉H₇F₃N₂O₃
MDL: MFCD00052846
CAS-Nummer: 395-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR