6-Chlor-5-nitronicotinsäure

Lieferant: COMBI-BLOCKS

PY-7038-1G PY-7038-5G
COBBPY-7038-1GEA 0 EUR
COBBPY-7038-1G COBBPY-7038-5G
6-Chlor-5-nitronicotinsäure
6-Chlor-5-nitronicotinsäure
Formel: C₆H₃ClN₂O₄
Molecular Weight: 202,5520 g/mol
MDL: MFCD09027297
CAS-Nummer: 7477-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR