Ethyl-2-aminonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-aminonicotinate

PY-7812-1G PY-7812-25G PY-7812-5G
COBBPY-7812-1GEA 26.2 EUR
COBBPY-7812-1G COBBPY-7812-25G COBBPY-7812-5G
Ethyl-2-aminonicotinat
Ethyl-2-aminonicotinat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.18 g/mol
MDL: MFCD01569452
CAS-Nummer: 13362-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR