Ethyl-2-aminoisonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: 2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate

CA-4047-5G CA-4047-1G
COBBCA-4047-5GEA 95.9 EUR
COBBCA-4047-5G COBBCA-4047-1G
Ethyl-2-aminoisonicotinat
Ethyl-2-aminoisonicotinat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166.18 g/mol
MDL: MFCD03791260
CAS-Nummer: 13362-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR