Ethyl-2-methylnicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-methylnicotinate

OS-7641-5G OS-7641-25G OS-7641-1G
COBBOS-7641-5GEA 50.9 EUR
COBBOS-7641-5G COBBOS-7641-25G COBBOS-7641-1G
Ethyl-2-methylnicotinat
Ethyl-2-methylnicotinat
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
MDL: MFCD00006336
CAS-Nummer: 1721-26-2
EINECS: 217-013-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR