Indium(III)chlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck

57110-10G 57110-50G
SIAL57110-10GEA 91.1 EUR
SIAL57110-10G SIAL57110-50G
Indium(III)chlorid Hydrat, Sigma-Aldrich®
Indium(III)chlorid Hydrat
Formel: Cl₃In·H₂O
Molecular Weight: 221.18 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149744
CAS-Nummer: 143983-91-9
EINECS: 233-043-0
UN: 3260
ADR: 8 II
Merck Index: 13,04980

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR