Indium(III)hydroxid ≥99,8% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40100.09
40100.18
40100.09EA
59.9
EUR
40100.09
40100.18
Indium(III)hydroxid ≥99,8% (Metall-Basis)
Indium(III) hydroxide
Purity: 99.8%
CAS No.: 20661-21-6
Molecular Formula: In(OH)3
Formula Weight: 165.84
Physical Form: Powder
Melting Point: 150° -H2O
MDL No.: MFCD00011056
Formel:
H₃InO₃ Molecular Weight: 165.84 g/mol Schmelzpunkt: 150 °C (-H₂O) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011056 CAS-Nummer: 20661-21-6 EINECS: 243-947-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...