Neodymnitrid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Scientific
Synonyme:
Neodymium(III) nitride
42091.04
42091.09
42091.04EA
73
EUR
42091.04
42091.09
Neodymnitrid ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Neodymium nitride
Purity: 99.9%
CAS No.: 25764-11-8
Molecular Formula: NdN
Formula Weight: 158.25
Physical Form: -60 Mesh Powder
Formel:
NNd Molecular Weight: 158,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00049990 CAS-Nummer: 25764-11-8 EINECS: 247-246-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...