Zirkonium(IV)sulfat Tetrahydrat ≥99,99% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

43194.22 43194.14
43194.22EA 784 EUR
43194.22 43194.14
Zirkonium(IV)sulfat Tetrahydrat ≥99,99% (Metall-Basis)
Zirkonium(IV)sulfat Tetrahydrat

Purity: 99.99% CAS No.: 7446-31-3 Molecular Formula: Zr(SO4)2.4H2O Formula Weight: 355.40 (283.34anhy) Physical Form: Crystalline powder Density: 3.22 Appearance: White Merck Reference: 14,10182 MDL No.: MFCD00149267

Formel: O₈S₂Zr·4H₂O
Molecular Weight: 355,396 g/mol
Dichte: 3,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00149267
CAS-Nummer: 7446-31-3
EINECS: 238-694-4
Merck Index: 14,10182

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR