Rhodium(III)oxid, wasserfrei ≥99,9% (Metall-Basis) Rh ≥ 80.6%

Lieferant: Thermo Scientific

11814.02 11814.04
11814.02EA 1036 EUR
11814.02 11814.04
Rhodium(III)oxid, wasserfrei ≥99,9% (Metall-Basis) Rh ≥ 80.6%
Rhodium(III) oxide

Purity: 99.9% CAS No.: 12036-35-0 Molecular Formula: Rh2O3 Formula Weight: 253.81 Physical Form: Powder Density: 8.20 Appearance: Grey to black Odour: Odourless Melting Point: 1100° dec. MDL No.: MFCD00011205

Formel: O₃Rh₂
Molecular Weight: 253,81 g/mol
Schmelzpunkt: 1100 °C (Zers.)
Dichte: 8,20
Lagertemperatur: Raumtemperatur
MDL: MFCD00011205
CAS-Nummer: 12036-35-0
EINECS: 234-846-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR