1-Brom-4-(brommethyl)naphthalin

Lieferant: COMBI-BLOCKS

CS-9099-1G CS-9099-5G
COBBCS-9099-1GEA 69 EUR
COBBCS-9099-1G COBBCS-9099-5G
1-Brom-4-(brommethyl)naphthalin
1-Bromo-4-(bromomethyl)naphthalene
Formel: C₁₁H₈Br₂
Molecular Weight: 299,9892 g/mol
MDL: MFCD07363811
CAS-Nummer: 79996-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR