3-(4-Methoxybenzoyl)propionsäure 98+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(4-Methoxyphenyl)-4-oxobutanoic acid

A18210.06 A18210.14
A18210.06EA 36.5 EUR
A18210.06 A18210.14
3-(4-Methoxybenzoyl)propionsäure 98+%
3-(4-Methoxybenzoyl)propionsäure

Purity: 98+% CAS No.: 3153-44-4 Molecular Formula: C11H12O4 Formula Weight: 208.21 Melting Point: 147-151° Beilstein Reference: 785074

Formel: C₁₁H₁₂O₄
Molecular Weight: 208.21 g/mol
Schmelzpunkt: 147…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002795
CAS-Nummer: 3153-44-4
EINECS: 221-593-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR