1,2-Diamino-9,10-anthrachinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Diaminoanthraquinone, 1,2-Diamino-10-anthracenedione

M-Clarity™ Qualitätsstufe = MQ100

D11582-100G D11582-25G D11582-5G
SIALD11582-100GEA 275 EUR
SIALD11582-100G SIALD11582-25G SIALD11582-5G
1,2-Diamino-9,10-anthrachinon, Sigma-Aldrich®
1,2-Diamino-9,10-anthrachinon
Formel: C₁₄H₁₀N₂O₂
Molecular Weight: 238,241 g/mol
MDL: MFCD00001219
CAS-Nummer: 1758-68-5
EINECS: 217-156-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR