2,6-Diamino-9,10-anthrachinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Diaminoanthracene-9,10-dione

M-Clarity™ Qualitätsstufe = MQ100

D12007-100G D12007-5G
SIALD12007-100GEA 130 EUR
SIALD12007-100G SIALD12007-5G
2,6-Diamino-9,10-anthrachinon, Sigma-Aldrich®
2,6-Diamino-9,10-anthrachinon
Formel: C₁₄H₁₀N₂O₂
Molecular Weight: 238,2414 g/mol
MDL: MFCD00001234
CAS-Nummer: 131-14-6
EINECS: 205-013-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR