5-Brom-4-isooctylpyrimidin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-4-(6-methylheptyl)pyrimidine

HC-6114-5G HC-6114-1G
COBBHC-6114-5GEA 451 EUR
COBBHC-6114-5G COBBHC-6114-1G
5-Brom-4-isooctylpyrimidin
5-Brom-4-isooctylpyrimidin
Formel: C₁₂H₁₉BrN₂
MDL: MFCD09800941
CAS-Nummer: 951884-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR