2-Chlor-2'-methoxyacetanilid

Lieferant: BIONET RESEARCH

Synonyme: 2-Chloro-N-(2-methoxyphenyl)acetamide

9G-909.500 9G-909.1 9G-909.5
BIOT9G-909.500EA 0 EUR
BIOT9G-909.500 BIOT9G-909.1 BIOT9G-909.5
2-Chlor-2'-methoxyacetanilid
2-Chlor-2'-methoxyacetanilid
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,6300 g/mol
MDL: MFCD00095394
CAS-Nummer: 55860-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR