2-Brom-6-fluor-3-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Bromo-6-fluoro-3-methylphenyl)boronic acid

BB-5009-1G BB-5009-5G
COBBBB-5009-1GEA 102 EUR
COBBBB-5009-1G COBBBB-5009-5G
2-Brom-6-fluor-3-methylphenylboronsäure
2-Brom-6-fluor-3-methylphenylboronsäure
Formel: C₇H₇BBrFO₂
Molecular Weight: 232,8400 g/mol
MDL: MFCD09800874
CAS-Nummer: 957121-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR