4-Chlor-6,7-dimethoxychinazolin
Lieferant: FLUOROCHEM
CFF97.1
017085-5G
017085-1G
CHMPCFF97.1EA
508
EUR
CHMPCFF97.1
FLUO017085-5G
FLUO017085-1G
4-Chlor-6,7-dimethoxychinazolin
4-Chloro-6,7-dimethoxyquinazoline
Formel:
C₁₀H₉ClN₂O₂ Molecular Weight: 224,65 g/mol |
MDL:
MFCD01570172 CAS-Nummer: 13790-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...