Diethylphthalat-[Ring-D4] (98% D)

Lieferant: LGC Standards PROMOCHEM

PROOCILDLM16290.25EA 954 EUR
PROOCILDLM16290.25 PROOCILDLM16290.1
Diethylphthalat-[Ring-D4] (98% D)
Diethylphthalat-[Ring-D4]
Formel: C₁₂H₁₀D₄O₄
MDL: MFCD00144117
CAS-Nummer: 93952-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR