1-Benzyl-4-hydroxypiperidin

Lieferant: FLUOROCHEM

Synonyme: 1-Benzylpiperidin-4-ol

023288-25G 023288-5G
FLUO023288-25GEA 57.9 EUR
FLUO023288-25G FLUO023288-5G
1-Benzyl-4-hydroxypiperidin
1-Benzyl-4-hydroxypiperidin
Formel: C₁₂H₁₇NO
Molecular Weight: 191,27 g/mol
MDL: MFCD00006503
CAS-Nummer: 4727-72-4
EINECS: 225-226-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR