4-Benzyl-4-hydroxypiperidin

Lieferant: Thermo Scientific

Synonyme: 4-Benzylpiperidin-4-ol

JFD00691.1 JFD00691.10
MAYBJFD00691.1EA 0 EUR
MAYBJFD00691.1 MAYBJFD00691.10
4-Benzyl-4-hydroxypiperidin
4-Benzyl-4-hydroxypiperidin
Formel: C₁₂H₁₇NO
Molecular Weight: 191,27 g/mol
MDL: MFCD00023147
CAS-Nummer: 51135-96-7
EINECS: 257-003-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR