2-Methoxy-4-nitrobenzonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

H27391.14 H27391.06
H27391.14EA 331 EUR
H27391.14 H27391.06
2-Methoxy-4-nitrobenzonitril ≥98%
2-Methoxy-4-nitrobenzonitril

Purity: 98% CAS No.: 101084-96-2 Molecular Formula: C8H6N2O3 Formula Weight: 178.15 Melting Point: 177-180° UN No.: 3439 MDL No.: MFCD02093781

Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
Schmelzpunkt: 177…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093781
CAS-Nummer: 101084-96-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR