2-Amino-N-methylbenzamid
Lieferant: FLUOROCHEM
018186-10G
018186-5G
FLUO018186-10GEA
63.9
EUR
FLUO018186-10G
FLUO018186-5G
2-Amino-N-methylbenzamid
2-Amino-N-methylbenzamid
Formel:
C₈H₁₀N₂O Molecular Weight: 150,1778 g/mol |
MDL:
MFCD00060602 CAS-Nummer: 4141-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...