4-Amino-1-isopropylpiperidin

Lieferant: CHEMPUR

60-0006.1 60-0006.5
CHMP60-0006.1EA 0 EUR
CHMP60-0006.1 CHMP60-0006.5
4-Amino-1-isopropylpiperidin
4-Amino-1-isopropylpiperidin
Formel: C₈H₁₈N₂
Molecular Weight: 142,2419 g/mol
MDL: MFCD03411606
CAS-Nummer: 127285-08-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR