4-Amino-1-methylpiperidin

Lieferant: CHEMPUR

Synonyme: 1-Methylpiperidin-4-amine

60-0153.1 60-0153.5
CHMP60-0153.1EA 0 EUR
CHMP60-0153.1 CHMP60-0153.5
4-Amino-1-methylpiperidin
4-Amino-1-methylpiperidin
Formel: C₆H₁₄N₂
Molecular Weight: 114.1888 g/mol
MDL: MFCD01570549
CAS-Nummer: 41838-46-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR