3-Methyl-2-thiophencarbonitril 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Methylthiophene-2-carbonitrile

A13305.06 A13305.03 A13305.14
A13305.06EA 98.4 EUR
A13305.06 A13305.03 A13305.14
3-Methyl-2-thiophencarbonitril 96%
3-Methyl-2-thiophencarbonitril

Purity: 96% CAS No.: 55406-13-8 Molecular Formula: C6H5NS Formula Weight: 123.18 Refractive Index: 1.5530 Boiling Point: 91-93°/12mm UN No.: 3276 MDL No.: MFCD00052086

Formel: C₆H₅NS
Molecular Weight: 123,18 g/mol
Siedepunkt: 91…93 °C (12 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052086
CAS-Nummer: 55406-13-8
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR