Monomethylphthalat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Methoxycarbonyl)benzoic acid, Methyl hydrogen phthalate

A14780.09 A14780.18 A14780.30 AB132028.0250
A14780.09EA 42.5 EUR
A14780.09 A14780.18 A14780.30 ABCRAB132028.0250
Monomethylphthalat ≥97%
Monomethylphthalat

Purity: 97% CAS No.: 4376-18-5 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 81-84° Beilstein Reference: 2048766

Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 81…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002466
CAS-Nummer: 4376-18-5
EINECS: 224-476-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR