6-Hydroxy-5-nitronicotinsäure
Lieferant: COMBI-BLOCKS
CA-4120-25G
CA-4120-1G
CA-4120-5G
COBBCA-4120-25GEA
0
EUR
COBBCA-4120-25G
COBBCA-4120-1G
COBBCA-4120-5G
6-Hydroxy-5-nitronicotinsäure
6-Hydroxy-5-nitronicotinsäure
Formel:
C₆H₄N₂O₅ |
MDL:
MFCD02180840 CAS-Nummer: 6635-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...