6-Brompiperonal

Lieferant: FLUOROCHEM

Synonyme: 6-Brom-1,3-benzodioxol-5-carbaldehyd

011589-10G 011589-100G
FLUO011589-10GEA 165 EUR
FLUO011589-10G FLUO011589-100G
6-Brompiperonal
6-Brompiperonal
Formel: C₈H₅BrO₃
Molecular Weight: 229.03 g/mol
MDL: MFCD00022952
CAS-Nummer: 15930-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR