2-Brom-5-chlorpyridin
Lieferant: FLUOROCHEM
012730-100G
012730-25G
FLUO012730-100GEA
200
EUR
FLUO012730-100G
FLUO012730-25G
2-Brom-5-chlorpyridin
2-Bromo-5-chloropyridine
Formel:
C₅H₃BrClN Molecular Weight: 192.44 g/mol |
MDL:
MFCD00234006 CAS-Nummer: 40473-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...