5-Amino-2-methylpyridin

Lieferant: FLUOROCHEM

Synonyme: 6-Methylpyridin-3-amin, 3-Amino-6-picolin, 6-Methyl-3-pyridylamin, 5-Amino-2-picolin

012721-1G 012721-5G
FLUO012721-1GEA 31.1 EUR
FLUO012721-1G FLUO012721-5G
5-Amino-2-methylpyridin
5-Amino-2-methylpyridin
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
MDL: MFCD00833389
CAS-Nummer: 3430-14-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR