2'-(Trifluormethyl)acetanilid

Lieferant: Thermo Scientific

SPB06386.1 SPB06386.10 SPB06386.50
MAYBSPB06386.1EA 0 EUR
MAYBSPB06386.1 MAYBSPB06386.10 MAYBSPB06386.50
2'-(Trifluormethyl)acetanilid
2'-(Trifluormethyl)acetanilid
Formel: C₉H₈F₃NO
MDL: MFCD00013555
CAS-Nummer: 344-62-7
EINECS: 206-453-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR