N,N-Dimethylformamiddimethylacetal

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 1,1-Dimethoxytrimethylamin

005030-500G 005030-100G
FLUO005030-500GEA 195 EUR
FLUO005030-500G FLUO005030-100G
N,N-Dimethylformamiddimethylacetal
N,N-Dimethylformamiddimethylacetal
Formel: C₅H₁₃NO₂
Molecular Weight: 119.16 g/mol
Siedepunkt: 102
Dichte: 0.893 g/cm³ (20 °C)
Flammpunkt: 6
MDL: MFCD00008482
CAS-Nummer: 4637-24-5
EINECS: 225-063-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR