3-(Trifluormethoxy)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3-(Trifluormethoxy)benzol-1-sulfonylchlorid, 3-(Trifluormethoxy)benzolsulfonsäurechlorid, 3-(Trifluormethoxy)benzolsulfochlorid
009000-1G
009000-5G
009000-25G
FLUO009000-1GEA
31.1
EUR
FLUO009000-1G
FLUO009000-5G
FLUO009000-25G
3-(Trifluormethoxy)benzolsulfonylchlorid
3-(Trifluormethoxy)benzolsulfonylchlorid
Formel:
C₇H₄ClF₃O₃S Molecular Weight: 260,62 g/mol Siedepunkt: 73…74 °C (0,7 mmHg) |
MDL:
MFCD01091016 CAS-Nummer: 220227-84-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...