3,4,5-Trifluorbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

013086-1G 013086-5G
FLUO013086-1GEA 31.1 EUR
FLUO013086-1G FLUO013086-5G
3,4,5-Trifluorbenzolsulfonylchlorid
3,4,5-Trifluorbenzolsulfonylchlorid
Formel: C₆H₂ClF₃O₂S
MDL: MFCD03094559
CAS-Nummer: 351003-43-5
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR