ω-Brom-2,4-dimethoxyacetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dimethoxyphenacylbromid, 2-Brom-2',4'-dimethoxyacetophenon, α-Brom-2,4-dimethoxyacetophenon, 2-Brom-1-(2,4-dimethoxyphenyl)ethan-1-on

018062-5G 018062-1G
FLUO018062-5GEA 0 EUR
FLUO018062-5G FLUO018062-1G
ω-Brom-2,4-dimethoxyacetophenon
ω-Brom-2,4-dimethoxyacetophenon
Formel: C₁₀H₁₁BrO₃
Molecular Weight: 259.1 g/mol
Siedepunkt: 315
Schmelzpunkt: 154…156 °C
MDL: MFCD00000197
CAS-Nummer: 60965-26-6
EINECS: 262-542-6
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR