2-Fluor-3-(trifluormethyl)benzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Fluoro-3-(trifluoromethyl)benzene-1-carbonyl chloride
006402-1G
006402-5G
006402-10G
FLUO006402-1GEA
73.1
EUR
FLUO006402-1G
FLUO006402-5G
FLUO006402-10G
2-Fluor-3-(trifluormethyl)benzoylchlorid
2-Fluor-3-(trifluormethyl)benzoylchlorid
Formel:
C₈H₃ClF₄O Molecular Weight: 226.56 g/mol Siedepunkt: 192…194 °C (1013 hPa) Dichte: 1.517 g/cm³ (20 °C) |
MDL:
MFCD00061154 CAS-Nummer: 208173-19-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...