5-Fluor-2-(trifluormethyl)benzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 5-Fluor-2-(trifluormethyl)benzol-1-carbonylchlorid, 5-Fluor-2-(trifluormethyl)benzoesäurechlorid

007918-5G 007918-25G
FLUO007918-5GEA 128 EUR
FLUO007918-5G FLUO007918-25G
5-Fluor-2-(trifluormethyl)benzoylchlorid
5-Fluor-2-(trifluormethyl)benzoylchlorid
Formel: C₈H₃ClF₄O
Molecular Weight: 226.56 g/mol
Siedepunkt: 168…169 °C (1013 hPa)
Dichte: 1.5 g/cm³ (20 °C)
MDL: MFCD00083534
CAS-Nummer: 216144-70-6
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR