Yttrium(III)acetylacetonat Hydrat ≥99,9% (Y-Basis)

Lieferant: Strem Chemicals, Inc.
Achtung

Synonyme: Tris(2,4-pentandionato-O,O')yttrium(III) Hydrat, Yttrium(III)-2,4-pentandionat Hydrat, Tri(2,4-pentandionato-O,O')yttrium(III) Hydrat, Yttrium(III)-tris(2,4-pentandionato-O,O') Hydrat

93-3901-100G AB122284.0005
STRE93-3901-100GEA 750 EUR
STRE93-3901-100G ABCRAB122284.0005
Yttrium(III)acetylacetonat Hydrat ≥99,9% (Y-Basis)
Yttrium(III)acetylacetonat Hydrat
Formel: C₁₅H₂₁O₆Y·H₂O
Molecular Weight: 404.25 g/mol
MDL: MFCD00013509
CAS-Nummer: 207801-29-4
EINECS: 239-607-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR