2-Fluor-4-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

006423-1G 006423-5G
FLUO006423-1GEA 134 EUR
FLUO006423-1G FLUO006423-5G
2-Fluor-4-(trifluormethyl)benzylbromid
2-Fluor-4-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Siedepunkt: 197
Schmelzpunkt: 22
MDL: MFCD00061173
CAS-Nummer: 239087-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR