2-Fluor-5-(trifluormethyl)benzylbromid
Lieferant: FLUOROCHEM
Synonyme:
1-(Brommethyl)-2-fluor-5-(trifluormethyl)benzol
006424-1G
006424-5G
FLUO006424-1GEA
36.9
EUR
FLUO006424-1G
FLUO006424-5G
2-Fluor-5-(trifluormethyl)benzylbromid
2-Fluor-5-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrF₄ Molecular Weight: 257.03 g/mol Siedepunkt: 197 Schmelzpunkt: 22 |
MDL:
MFCD00061174 CAS-Nummer: 220239-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...