2-Fluor-6-(trifluormethyl)benzylbromid
Lieferant: FLUOROCHEM
Synonyme:
1-(Brommethyl)-2-fluor-6-(trifluormethyl)benzol
006425-1G
006425-5G
FLUO006425-1GEA
36.9
EUR
FLUO006425-1G
FLUO006425-5G
2-Fluor-6-(trifluormethyl)benzylbromid
2-Fluor-6-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrF₄ Molecular Weight: 257.03 g/mol Siedepunkt: 197 Schmelzpunkt: 43…45 °C |
MDL:
MFCD00082477 CAS-Nummer: 239087-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...