3-Fluor-4-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-(Brommethyl)-3-fluor-4-(trifluormethyl)benzol

007924-1G 007924-5G
FLUO007924-1GEA 106 EUR
FLUO007924-1G FLUO007924-5G
3-Fluor-4-(trifluormethyl)benzylbromid
3-Fluor-4-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Siedepunkt: 197
Schmelzpunkt: 22
MDL: MFCD00236289
CAS-Nummer: 213203-65-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR