5-Fluor-2-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

007925-5G 007925-10G
FLUO007925-5GEA 60.9 EUR
FLUO007925-5G FLUO007925-10G
5-Fluor-2-(trifluormethyl)benzylbromid
5-Fluor-2-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Siedepunkt: 197
Schmelzpunkt: 22
MDL: MFCD00236290
CAS-Nummer: 239135-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR