4-Fluor-2-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-(Brommethyl)-4-fluor-2-(trifluormethyl)benzol

006427-1G 006427-5G 006427-25G
FLUO006427-1GEA 34.1 EUR
FLUO006427-1G FLUO006427-5G FLUO006427-25G
4-Fluor-2-(trifluormethyl)benzylbromid
4-Fluor-2-(trifluormethyl)benzylbromid
Formel: C₈H₅BrF₄
Molecular Weight: 257.03 g/mol
Siedepunkt: 197
Schmelzpunkt: 22
MDL: MFCD00061176
CAS-Nummer: 206860-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR