3-Chlor-4-fluor-5-nitrobenzotrifluorid
Lieferant: FLUOROCHEM
Synonyme:
1-Chloro-2-fluoro-3-nitro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluoro-5-nitrobenzotrifluoride
009085-1G
009085-5G
FLUO009085-1GEA
0
EUR
FLUO009085-1G
FLUO009085-5G
3-Chlor-4-fluor-5-nitrobenzotrifluorid
3-Chlor-4-fluor-5-nitrobenzotrifluorid
Formel:
C₇H₂ClF₄NO₂ Molecular Weight: 243.54 g/mol |
MDL:
MFCD00674108 CAS-Nummer: 101646-02-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...