2,4-Dibromthiazol

Lieferant: FLUOROCHEM

Synonyme: 2,4-dibromo-1,3-thiazole

016251-1G 016251-25G 016251-5G
FLUO016251-1GEA 31.1 EUR
FLUO016251-1G FLUO016251-25G FLUO016251-5G
2,4-Dibromthiazol
2,4-Dibromothiazole
Formel: C₃HBr₂NS
Molecular Weight: 242.92 g/mol
MDL: MFCD01318994
CAS-Nummer: 4175-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR