2-Brom-3-(trifluormethyl)pyridin
Lieferant: FLUOROCHEM
012722-1G
012722-5G
FLUO012722-1GEA
31.1
EUR
FLUO012722-1G
FLUO012722-5G
2-Brom-3-(trifluormethyl)pyridin
2-Bromo-3-(trifluoromethyl)pyridine
Formel:
C₆H₃BrF₃N Molecular Weight: 226 g/mol |
MDL:
MFCD00153084 CAS-Nummer: 175205-82-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...